Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DNA Ligase IV antibody (YA2699) | 10 UL
SDP

Catalog No. 5000445136
Encompass_Preferred

DNA Ligase IV antibody (YA2699) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets DNA Ligase IV. This antibody plays a critical role in DNA non-homologous end joining (NHEJ), specifically in double-strand break (DSB) repair and V(D)J recombination, by catalyzing the NHEJ ligation step to reseal DNA breaks.

  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Targets DNA Ligase IV
  • Reactivity: Human
  • Application: WB (Western Blot)
  • Dilution ratio: 1:500-1:1000 for WB
  • SwissProt ID: P49917
  • Research field: Epigenetics and nuclear signaling
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Appearance: Liquid
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-45136 Supplier Medchemexpress LLC Supplier No. HYP8295410UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA Ligase IV antibody (YA2699) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets DNA Ligase IV. This antibody plays a critical role in DNA non-homologous end joining (NHEJ), specifically in double-strand break (DSB) repair and V(D)J recombination, by catalyzing the NHEJ ligation step to reseal DNA breaks.

  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Targets DNA Ligase IV
  • Reactivity: Human
  • Application: WB (Western Blot)
  • Dilution ratio: 1:500-1:1000 for WB
  • SwissProt ID: P49917
  • Research field: Epigenetics and nuclear signaling
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Appearance: Liquid
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.