Learn More
Sigma Aldrich Fine Chemicals Biosciences DNA Ligation Kit
Catalog No. 502904283
Encompass
Sigmas DNA Ligation Kit contains all of the reagents necessary to perform DNA ligation reactions. Sticky-end ligations are more efficient than blunt-end ligations these can be facilitated with the addition of PEG.
Catalog No. 50-290-4283 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. LIG11KT
Sigmas DNA Ligation Kit contains all of the reagents necessary to perform DNA ligation reactions. Sticky-end ligations are more efficient than blunt-end ligations these can be facilitated with the addition of PEG.