Learn More
Sigma Aldrich Fine Chemicals Biosciences DNA MB grade from fish spe
Catalog No. 502906122
Encompass
DNA MB-grade is provided as a sonicated denatured single-stranded DNA fragment mixture.
Catalog No. 50-290-6122 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11467140001
DNA MB-grade is provided as a sonicated denatured single-stranded DNA fragment mixture.