Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical DNA MeC(C6-O-Lev) amidite | 5 g | CAS 135887-12-6 | MDL MFCD15145422

Catalog No. 502362749
Encompass

DNA MeC(C6-O-Lev) amidite is a Amidite reagent (Subcategory: Cytosine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 135887-12-6
- MDL: MFCD15145422
- InChIKey: YIGBFFPGFGSLIZ-VCTOYPRMSA-N
- Molecular Weight: 942.103761998
- Molecular Formula: C51H68N5O10P
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 6-((1-((2R,4S,5R)-5-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)tetrahydrofuran-2-yl)-5-methyl-2-oxo-1,2-dihydropyrimidin-4-yl)amino)hexyl 4-oxopentanoate
- SMILES: CC(N(P(O[C@H]1C[C@H](N2C(N=C(C(C)=C2)NCCCCCCOC(CCC(C)=O)=O)=O)O[C@@H]1COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OCCC#N)C(C)C)C

Catalog No. 50-236-2749 Supplier STA Pharmaceutical Supplier No. STAAM10653AK1005GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA MeC(C6-O-Lev) amidite is a Amidite reagent (Subcategory: Cytosine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 135887-12-6
- MDL: MFCD15145422
- InChIKey: YIGBFFPGFGSLIZ-VCTOYPRMSA-N
- Molecular Weight: 942.103761998
- Molecular Formula: C51H68N5O10P
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 6-((1-((2R,4S,5R)-5-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)tetrahydrofuran-2-yl)-5-methyl-2-oxo-1,2-dihydropyrimidin-4-yl)amino)hexyl 4-oxopentanoate
- SMILES: CC(N(P(O[C@H]1C[C@H](N2C(N=C(C(C)=C2)NCCCCCCOC(CCC(C)=O)=O)=O)O[C@@H]1COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.