Learn More
Enzo Life Sciences Express DNA methylation kit (shallow well) (2x96wells)
SDP
Catalog No. 502012564
Encompass_Preferred
The Express DNA methylation kit (shallow well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis. No preparation is necessary when using the Express Conversion Reagent. Simply add this reagent to a DNA sample and let the reaction proceed to completion. DNA denaturation and bisulfite conversion processes are combined with added heat to facilitate rapid denaturation. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Catalog No. 50-201-2564 Supplier Enzo Life Sciences Supplier No. ENZ450030002
The Express DNA methylation kit (shallow well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis. No preparation is necessary when using the Express Conversion Reagent. Simply add this reagent to a DNA sample and let the reaction proceed to completion. DNA denaturation and bisulfite conversion processes are combined with added heat to facilitate rapid denaturation. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.