Learn More
Cayman Chemical DNA MethyltrnsfRase 3a Mon 1ea
SDP
Catalog No. 502955238
Encompass_Preferred
Antigen: His-tag recombinant mouse Dnmt3a expressed in bacteria • Isotype: IgG1 • Host: mouse, clone 64B814.1 • Cross Reactivity: (+) human and mouse DNMT3a • Application(s): ELISA • The DNMT3 family members DNMT3a and DNMT3b are strongly expressed in embryonic stem cells. Their expression is down regulated in differentiating cells and low in adult sematic tissue.
Catalog No. 50-295-5238 Supplier Cayman Chemical Supplier No. 134831 EA
Antigen: His-tag recombinant mouse Dnmt3a expressed in bacteria • Isotype: IgG1 • Host: mouse, clone 64B814.1 • Cross Reactivity: (+) human and mouse DNMT3a • Application(s): ELISA • The DNMT3 family members DNMT3a and DNMT3b are strongly expressed in embryonic stem cells. Their expression is down regulated in differentiating cells and low in adult sematic tissue.