Learn More
Enzymax LLC DNA MIDI SPIN COLUMN 25/PK
SDP
Catalog No. NC1405068
Encompass
DNA Midi spin column 25/pkDNA midi Spin columns offer a simple, rapid, and cost effective method for DNA isolation from various samples in medium scale. They can be used for different applications by using different buffers and protocols. Like Mini and Tini spin columns, Midi columns contain a silica-based membrane and they are compatible with all the buffers that are used for Mini and Tini columns. These columns can be used with both centrifuge and vacuum manifold. Midi filter column can be connected to Midi spin column for cell lysate clarification by using vacuum manifold.Collection tubes are included.
Catalog No. NC1405068 Supplier Enzymax LLC Supplier No. EZC222
DNA Midi spin column 25/pkDNA midi Spin columns offer a simple, rapid, and cost effective method for DNA isolation from various samples in medium scale. They can be used for different applications by using different buffers and protocols. Like Mini and Tini spin columns, Midi columns contain a silica-based membrane and they are compatible with all the buffers that are used for Mini and Tini columns. These columns can be used with both centrifuge and vacuum manifold. Midi filter column can be connected to Midi spin column for cell lysate clarification by using vacuum manifold.Collection tubes are included.