Learn More
Sigma Aldrich Fine Chemicals Biosciences DNA Molecular Weight Marke
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11669940910
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical. The DIG System was established as a sensitive and cost-effective alternative to using radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.DNA Molecular Weight Marker VII DIG-labeled is a ready-to-use SOLUTIONon in 10mM Tris-HCl 1mM EDTA pH 8.0. The fragment mixture is prepared by cleavage of SPP1 DNA with EcoRI. the size ranges from 0.081 to 8.57kbp.
Catalog No. 50-290-6218