Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical DNA N3-Me-T amidite | 5 g | CAS 630120-88-6 | InChIKey YNUFSWZIKOXYIY-VPJZQFIISA-N

Catalog No. 502363857
Encompass

DNA N3-Me-T amidite is a Amidite reagent (Subcategory: Thymine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 630120-88-6
- MDL: No data
- InChIKey: YNUFSWZIKOXYIY-VPJZQFIISA-N
- Molecular Weight: 758.852761998
- Molecular Formula: C41H51N4O8P
- Purity: ≥98%
- Container Type: 250 mL Glass (28-400)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 174.7 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(3,5-dimethyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N(C(C(C)=CN1[C@H]2C[C@@H]([C@H](O2)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OP(N(C(C)C)C(C)C)OCCC#N)=O)C

Catalog No. 50-236-3857 Supplier STA Pharmaceutical Supplier No. STAAM10604MM2505GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA N3-Me-T amidite is a Amidite reagent (Subcategory: Thymine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 630120-88-6
- MDL: No data
- InChIKey: YNUFSWZIKOXYIY-VPJZQFIISA-N
- Molecular Weight: 758.852761998
- Molecular Formula: C41H51N4O8P
- Purity: ≥98%
- Container Type: 250 mL Glass (28-400)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 174.7 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(3,5-dimethyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N(C(C(C)=CN1[C@H]2C[C@@H]([C@H](O2)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OP(N(C(C)C)C(C)C)OCCC#N)=O)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.