Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical DNA N6-Etheno-A amidite | 1 g | CAS 160702-08-9 | InChIKey BWUNNFYWTPVXCM-GHFFOPBTSA-N

Catalog No. 502362953
Encompass

DNA N6-Etheno-A amidite is a Amidite reagent (Subcategory: Adenine Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 160702-08-9
- MDL: No data
- InChIKey: BWUNNFYWTPVXCM-GHFFOPBTSA-N
- Molecular Weight: 777.862761998
- Molecular Formula: C42H48N7O6P
- Purity: ≥95%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(3H-imidazo[2,1-i]purin-3-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=NC6=C5N=CN7C=CN=C67)C(C)C)C

Catalog No. 50-236-2953 Supplier STA Pharmaceutical Supplier No. STAAM10673MM301GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA N6-Etheno-A amidite is a Amidite reagent (Subcategory: Adenine Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 160702-08-9
- MDL: No data
- InChIKey: BWUNNFYWTPVXCM-GHFFOPBTSA-N
- Molecular Weight: 777.862761998
- Molecular Formula: C42H48N7O6P
- Purity: ≥95%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(3H-imidazo[2,1-i]purin-3-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=NC6=C5N=CN7C=CN=C67)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.