Learn More
Andwin Scientific DNA Off Decontaminant , 8.5oz
SDP
Supplier:  Andwin Scientific EF12982
Encompass_Preferred
DNA Off is a decontaminant that eliminates DNA more efficiently than autoclaving. This product has a capacity of 8.5 ounces.
Catalog No. NC9816121