Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA Paternity Testing Simulation Kit
SDPSCI_ED

Catalog No. S11114 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11114 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11114 Supplier Edvotek™ Supplier No. 114
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets.

Requires

  • Required Materials (not included):
  • Horizontal Gel Electrophoresis apparatus
  • Water bath
  • Visualization system (white light)
  • Micropipets
  • Hot-hand protector
  • Deionized water
  • 250mL beakers
  • Hotplate or Bunsen burner
  • Photodocumentation system

This experiment introduces students to the use of DNA fingerprinting in a simulated paternity determination. A child's DNA fingerprint is compared with his parents. The experiment does not contain human DNA.

This experiment introduces students to the use of DNA fingerprinting in a hypothetical paternity determination. A child's DNA fingerprint is compared with that of his parents. The experiment does not contain human DNA.

Specifications

Grade 9 to Undergraduate
Includes Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, and Disposable Pipets.
Quantity 8 Gels
Time Range Complete in 45 minutes
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.