Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ DNA Paternity Testing Simulation with SYBR™ Safe
SDPSCI_ED

Catalog No. 01672272 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
01-672-272 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-272 Supplier EDVOTEK™ Supplier No. 114S
Add to Cart
Add to Cart

This DNA Paternity Testing Simulation with SYBR™ Safe experiment introduces students to the use of DNA fingerprinting in a simulated paternity determination. A child's DNA fingerprint is compared with his parents.

  • Includes SYBR™ Safe DNA Stain, NOT FlashBlue™ DNA Stain
  • Learn genetics and molecular biology techniques by solving a hypothetical paternity case
  • Introduce students to restriction enzymes, PCR, and electrophoresis
  • Perform agarose gel electrophoresis to separate differently-sized DNA molecules
  • Load, run, analyze, and size pre-digested PCR samples to determine the biological parents of a child
  • Group Size: For 8 Gels
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe DNA Stain, Practice Gel Loading Solution.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade 8 to Undergraduate
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.