Learn More
ABclonal Technology DNA-PKcs Rabbit pAb
SDP
Catalog No. 5000107095
Encompass_Preferred
This gene encodes the catalytic subunit of the DNA-dependent protein kinase (DNA-PK). It functions with the Ku70/Ku80 heterodimer protein in DNA double strand break repair and recombination. The protein encoded is a member of the PI3/PI4-kinase family.
Catalog No. 50-001-07095 Supplier ABclonal Technology Supplier No. A14191000UL
This gene encodes the catalytic subunit of the DNA-dependent protein kinase (DNA-PK). It functions with the Ku70/Ku80 heterodimer protein in DNA double strand break repair and recombination. The protein encoded is a member of the PI3/PI4-kinase family.