Learn More
New England Biolabs, Inc. DNA Polymerase I, Large (Klenow) Fragment – 1000 units
SDP
Catalog No. 50811622
Encompass
DNA Polymerase I, Large (Klenow) Fragment is a proteolytic product of E. coli DNA Polymerase I which retains polymerization and 3' to 5' exonuclease activity, but has lost 5' to 3' exonuclease activity. Klenow retains the polymerization fidelity of the holoenzyme without degrading 5' termini.
Catalog No. 50-811-622 Supplier New England Biolabs, Inc. Supplier No. M0210M
DNA Polymerase I, Large (Klenow) Fragment is a proteolytic product of E. coli DNA Polymerase I which retains polymerization and 3' to 5' exonuclease activity, but has lost 5' to 3' exonuclease activity. Klenow retains the polymerization fidelity of the holoenzyme without degrading 5' termini.