Learn More
New England Biolabs, Inc. DNA Polymerase I (E. coli) – 500 units
SDP
Catalog No. 50811620
Encompass
DNA Polymerase I (E coli) is a DNA-dependent DNA polymerase with inherent 3' to 5' and 5' to 3' exonuclease activities. The 5' to 3' exonuclease activity removes nucleotides ahead of the growing DNA chain, allowing nick-translation.
Catalog No. 50-811-620 Supplier New England Biolabs, Inc. Supplier No. M0209S
DNA Polymerase I (E coli) is a DNA-dependent DNA polymerase with inherent 3' to 5' and 5' to 3' exonuclease activities. The 5' to 3' exonuclease activity removes nucleotides ahead of the growing DNA chain, allowing nick-translation.