Learn More
New England Biolabs, Inc. DNA Polymerase I, Large (Klenow) Fragment – 200 units
SDP
Catalog No. 50811623
Encompass
DNA Polymerase I, Large (Klenow) Fragment is a proteolytic product of E. coli DNA Polymerase I which retains polymerization and 3' to 5' exonuclease activity, but has lost 5' to 3' exonuclease activity. Klenow retains the polymerization fidelity of the holoenzyme without degrading 5' termini.
Catalog No. 50-811-623 Supplier New England Biolabs, Inc. Supplier No. M0210S
DNA Polymerase I, Large (Klenow) Fragment is a proteolytic product of E. coli DNA Polymerase I which retains polymerization and 3' to 5' exonuclease activity, but has lost 5' to 3' exonuclease activity. Klenow retains the polymerization fidelity of the holoenzyme without degrading 5' termini.