Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DNA polymerase beta Protein, Human (His) | 100 UG
SDP

Catalog No. 5000445270
Encompass_Preferred

DNA polymerase beta Protein, Human (His) is a DNA repair enzyme crucial for maintaining genomic integrity. It plays a vital role in base excision repair (BER), a fundamental pathway essential for DNA maintenance, replication, recombination, and drug resistance. This protein belongs to the DNA polymerase type-X family and is involved in multiple DNA repair mechanisms.

  • Involved in various types of DNA repair mechanisms
  • Performs base excision repair (BER)
  • Essential for DNA maintenance, replication, recombination, and drug resistance
  • Belongs to the DNA polymerase type-X family
  • Composed of an 8-kDa domain and a 31-kDa domain
  • Also involved in Nucleotide Excision Repair (NER), Mismatch Repair (MMR), and Double Strand Break Repair (DSBR)
  • Species: Human
  • Source: E. coli
  • Purity: ≥ 95%

Catalog No. 50-004-45270 Supplier Medchemexpress LLC Supplier No. HYP83633100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA polymerase beta Protein, Human (His) is a DNA repair enzyme crucial for maintaining genomic integrity. It plays a vital role in base excision repair (BER), a fundamental pathway essential for DNA maintenance, replication, recombination, and drug resistance. This protein belongs to the DNA polymerase type-X family and is involved in multiple DNA repair mechanisms.

  • Involved in various types of DNA repair mechanisms
  • Performs base excision repair (BER)
  • Essential for DNA maintenance, replication, recombination, and drug resistance
  • Belongs to the DNA polymerase type-X family
  • Composed of an 8-kDa domain and a 31-kDa domain
  • Also involved in Nucleotide Excision Repair (NER), Mismatch Repair (MMR), and Double Strand Break Repair (DSBR)
  • Species: Human
  • Source: E. coli
  • Purity: ≥ 95%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.