Learn More
Qiagen Beverly Inc DNA POLYMERASE I
Catalog No. 50305911
Encompass
DNA Polymerase; DNA Polymerase I; Conc. 10,000 U/ml; 5,000 U; incl. 10X buffer
Catalog No. 50-305-911 Supplier Qiagen Beverly Inc Supplier No. P7050L
DNA Polymerase; DNA Polymerase I; Conc. 10,000 U/ml; 5,000 U; incl. 10X buffer