Learn More
TARGETMOL CHEMICALS INC DNA polymerase-IN-1 1MG
SDP
Catalog No. 502876806
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DNA polymerase-IN-1, a DNA polymerase inhibitor, exhibited antiproliferative activity against tumor cells with a half-inhibitory concentration (IC50) of 20.7 uM. DNA polymerase-IN-1 inhibited Taq DNA polymerase with an IC50 of less than 250 uM. DNA polymerase-IN-1 was shown to inhibit Taq DNA polymerase by RT- DNA polymerase-IN-1 showed antiretroviral activity in vitro by RT-PCR assay and can be used to study oxidative damage and colorectal cancer. Purity 99%
Catalog No. 50-287-6806 Supplier TARGETMOL CHEMICALS INC Supplier No. T806441MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DNA polymerase-IN-1, a DNA polymerase inhibitor, exhibited antiproliferative activity against tumor cells with a half-inhibitory concentration (IC50) of 20.7 uM. DNA polymerase-IN-1 inhibited Taq DNA polymerase with an IC50 of less than 250 uM. DNA polymerase-IN-1 was shown to inhibit Taq DNA polymerase by RT- DNA polymerase-IN-1 showed antiretroviral activity in vitro by RT-PCR assay and can be used to study oxidative damage and colorectal cancer. Purity 99%