Learn More
Agilent Technologies DNA POLYMERSAE HERCULASE II
Catalog No. NC9390548
Encompass
II Fusion DNA Polymerase; Enhanced processivity; Inclusion of our exclusive ArchaeMaxx; Herculase;; 5X Herculase; II Reaction Buffer 1mL; Dimethyl Sulfoxide (DMSO) 1mL; 40 rxn; Storage: -20 deg.C
Catalog No. NC9390548 Supplier Agilent Technologies Supplier No. 600675
II Fusion DNA Polymerase; Enhanced processivity; Inclusion of our exclusive ArchaeMaxx; Herculase;; 5X Herculase; II Reaction Buffer 1mL; Dimethyl Sulfoxide (DMSO) 1mL; 40 rxn; Storage: -20 deg.C