Learn More
Biotage Plasmid MegaPrep kit, 16 samples, yield up to 5 mg, endotoxin level <0.1 EU/μg, supercoiled DNA, elution volume 18 mL, A260/A280 purity >1.8, A260/A230 purity >2.0, compatible with Biotage PhyPrep system
SDP
Supplier:  Biotage DPM161072KIT
Encompass
Plasmid MegaPrep kit, 16 samples, yield up to 5 mg, endotoxin level <0.1 EU/μg, supercoiled DNA, elution volume 18 mL, A260/A280 purity >1.8, A260/A230 purity >2.0, compatible with Biotage PhyPrep system
Catalog No. NC2153330