Learn More
Agilent Technologies DNA REAGENTS
Catalog No. NC2030928
Encompass
  • Designed for precise separation and quantification of low-concentration double-stranded DNA samples.
  • Compatible with Agilent 2100 Bioanalyzer system.
  • Suitable for quality control of low input Next Generation Sequencing libraries.
  • Enables DNA fragment analysis down to 5 pg/µl.
  • Complex DNA analysis sensitivity at 100 pg/µl.
  • Supports up to 10 chips per kit.
  • Includes high sensitivity DNA ladder, dye concentrate, and gel matrix.
  • Quick analysis run time of 45 minutes.
  • Stable for 4 months.
  • For research use only.
Catalog No. NC2030928 Supplier Agilent Technologies Supplier No. 50674627
  • Designed for precise separation and quantification of low-concentration double-stranded DNA samples.
  • Compatible with Agilent 2100 Bioanalyzer system.
  • Suitable for quality control of low input Next Generation Sequencing libraries.
  • Enables DNA fragment analysis down to 5 pg/µl.
  • Complex DNA analysis sensitivity at 100 pg/µl.
  • Supports up to 10 chips per kit.
  • Includes high sensitivity DNA ladder, dye concentrate, and gel matrix.
  • Quick analysis run time of 45 minutes.
  • Stable for 4 months.
  • For research use only.