Learn More
G-Biosciences DNA/RNA Conserve Collection Tube
SCI_ED
Catalog No. S04230
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
S04230 50/Pk.
1 options
Catalog No. S04230 Supplier G-Biosciences Supplier No. 7862182
Add to Cart
Add to Cart
DNA/RNA-Conserve is designed for the preservation of DNA/RNA in biological samples.
DNA/RNA conserve collection tube is supplied with 1 mL DNA/RNA conserve solution. DNA/RNA-Conserve is designed for the preservation of DNA/RNA in biological samples, including but not limited to; cells, mammalian tissues, bacteria, DNA/RNA-virus, yeast, plant tissues, biological fluids, field and test site collected specimens, swab kits, and so forth. Purified DNA/RNA when stored in Conserve solution is protected from degradation and will remain intact during prolonged storage. This formulation is a solution of non-toxic salts and preserving agents, optimized to preserve the integrity of purified DNA/RNA. Inactivation of DNases/RNases eliminates the urgency for immediate processing of the biological samples or cryopreservation. Samples may ne stored or transported without degradation of RNA samples.
  • DNA/RNA stabilization solution that permeates cells and tissues and inactivate the DNases/RNAses.
  • Convenient: No need to immediately freeze or process sample for DNA and RNA extraction. Simply add Conserve Solution and store the sample.
  • Stable: Tissues and cells stored in the solution are stable for 1 day at 37°C, for 1 week at room temperature (approx. 25°C), one month at 4°C and indefinitely at -20°C.
  • Mobility: Easy to carry and store field samples
  • Compatible: with various DNA/RNA extraction methods and kits.
Applications:
DNA/RNA-Conserve Solution is designed to store and preserve DNA and RNA in various samples such as cells, tissue, plant, soil, blood etc before RNA extraction without the need to freeze samples. DNA/RNA extracted from samples stored in Conserve Solution is compatible with downstream applications such as PCR, RT-PCR, northern blot, Sequencing and other downstream applications.
Specifications
Product Type Specimen Collection and Transport Tube
Includes 1 mL DNA/RNA Conserve Solution
Quantity 50/Pk.