Learn More
Alkali Scientific DNA & RNA Elution Buffer (EB Buffer) for PCR, Plasmid & Spin Column Cleanup, 250mL
SDP
Catalog No. 5000662357
Encompass_Preferred
DNA and RNA Elution Buffer is used to recover purified nucleic acids from spin column-based extraction systems and related purification methods. The solution is formulated to provide optimal conditions for releasing DNA and RNA while maintaining their structural integrity. It supports downstream applications such as PCR, sequencing, and cloning by minimizing inhibitors and preserving nucleic acid stability. The buffer is suitable for a wide range of molecular biology workflows including plasmid purification, PCR product cleanup, and genomic DNA or RNA extraction. It is commonly used in research, clinical, and biotechnology laboratories where consistent and efficient nucleic acid recovery is essential.
Catalog No. 50-006-62357 Supplier Alkali Scientific Supplier No. EB0250
DNA and RNA Elution Buffer is used to recover purified nucleic acids from spin column-based extraction systems and related purification methods. The solution is formulated to provide optimal conditions for releasing DNA and RNA while maintaining their structural integrity. It supports downstream applications such as PCR, sequencing, and cloning by minimizing inhibitors and preserving nucleic acid stability. The buffer is suitable for a wide range of molecular biology workflows including plasmid purification, PCR product cleanup, and genomic DNA or RNA extraction. It is commonly used in research, clinical, and biotechnology laboratories where consistent and efficient nucleic acid recovery is essential.