Learn More
Alkali Scientific DNA & RNA Elution Buffer (EB Buffer) for PCR, Plasmid & Spin Column Cleanup, 500mL
SDP
Catalog No. 5000662015
Encompass_Preferred
DNA & RNA Elution Buffer is a high-purity solution designed for efficient recovery of nucleic acids during purification workflows. It is commonly used in spin column-based extraction kits for eluting DNA and RNA from silica membranes after washing steps. The buffer is formulated to be PCR compatible, ensuring that eluted nucleic acids can be directly used in downstream applications such as PCR amplification, qPCR, sequencing, and cloning without additional cleanup. Its low ionic strength helps maximize recovery while maintaining nucleic acid stability. This buffer is widely used in molecular biology laboratories for plasmid purification, genomic DNA extraction, and RNA isolation procedures where reliable and contaminant-free elution is required.
Catalog No. 50-006-62015 Supplier Alkali Scientific Supplier No. EB0500
DNA & RNA Elution Buffer is a high-purity solution designed for efficient recovery of nucleic acids during purification workflows. It is commonly used in spin column-based extraction kits for eluting DNA and RNA from silica membranes after washing steps. The buffer is formulated to be PCR compatible, ensuring that eluted nucleic acids can be directly used in downstream applications such as PCR amplification, qPCR, sequencing, and cloning without additional cleanup. Its low ionic strength helps maximize recovery while maintaining nucleic acid stability. This buffer is widely used in molecular biology laboratories for plasmid purification, genomic DNA extraction, and RNA isolation procedures where reliable and contaminant-free elution is required.