Learn More
Bioventures Inc DNA/RNA ISOLATION
Supplier:  Bioventures Inc GRT
Encompass
MAPMARKER, 50-1000BP, TMR Labeled (50, 75, 100, 125, 150, 200, 250, 300, 350, 400, 450, 475, 500, 550, 600, 650, 700, 750, 800, 850, 900, 950, 1000bp), 8fmoles/band/ul Concentration
Catalog No. 50-272-366