Learn More
Zymo Research Corporation Direct-zol™ RNA MiniPrep Plus (50 Preps) (Product Supplied w/ 50 ml TRI Reagent™)
SDP
Catalog No. NC1761673
Encompass_Preferred
Upon lysis of the sample with TRIzol or similar, RNA and DNA is bound directly to the Zymo-Spin™ Column. Then simply spin, wash, and elute high-quality RNA and DNA into separate fractions. No phase separation, precipitation, or post purification steps are necessary. The eluted nucleic acids are suitable for all subsequent molecular manipulations and analyses including Next-Gen sequencing, RT/qPCR, hybridization, etc.
Catalog No. NC1761673 Supplier Zymo Research Corporation Supplier No. R2081
Upon lysis of the sample with TRIzol or similar, RNA and DNA is bound directly to the Zymo-Spin™ Column. Then simply spin, wash, and elute high-quality RNA and DNA into separate fractions. No phase separation, precipitation, or post purification steps are necessary. The eluted nucleic acids are suitable for all subsequent molecular manipulations and analyses including Next-Gen sequencing, RT/qPCR, hybridization, etc.