Learn More
Zymo Research Corporation DNA/RNA Shield, 250 ml
SDP
Catalog No. 501251706
Encompass_Preferred
DNA/RNA Shield reagent preserves the genetic integrity and expression profiles of samples at ambient temperatures and completely inactivates infectious agents (viruses, bacteria, fungi, & parasites). The unique product also prevents degradation from freeze-thaw cycling and unexpected freezer failures.
Catalog No. 50-125-1706 Supplier Zymo Research Corporation Supplier No. R1100250
DNA/RNA Shield reagent preserves the genetic integrity and expression profiles of samples at ambient temperatures and completely inactivates infectious agents (viruses, bacteria, fungi, & parasites). The unique product also prevents degradation from freeze-thaw cycling and unexpected freezer failures.