Learn More
Zymo Research Corporation DNA/RNA Shield (25 ml) (2X Concentrate)
SDP
Catalog No. 501251707
Encompass_Preferred
DNA/RNA Shield reagent preserves the genetic integrity and expression profiles of samples at ambient temperatures and completely inactivates infectious agents (viruses, bacteria, fungi, & parasites). The unique product also prevents degradation from freeze-thaw cycling and unexpected freezer failures.
Catalog No. 50-125-1707 Supplier Zymo Research Corporation Supplier No. R120025
DNA/RNA Shield reagent preserves the genetic integrity and expression profiles of samples at ambient temperatures and completely inactivates infectious agents (viruses, bacteria, fungi, & parasites). The unique product also prevents degradation from freeze-thaw cycling and unexpected freezer failures.