Learn More
Zymo Research Corporation DNA/RNA Shield Collection Tube (50 pack)
SDP
Catalog No. 501251696
Encompass_Preferred
DNA/RNA Shield Lysis and Collection tubes are designed for the streamlined collection, transport, and processing of biological specimen. They are pre-filled with DNA/RNA Shield and a lysis matrix tailored for tough-to-lyse- sample inputs from tissue to microbial samples. Collected samples do not require additional handling for processing and are ready for homogenization.
Catalog No. 50-125-1696 Supplier Zymo Research Corporation Supplier No. R1102
DNA/RNA Shield Lysis and Collection tubes are designed for the streamlined collection, transport, and processing of biological specimen. They are pre-filled with DNA/RNA Shield and a lysis matrix tailored for tough-to-lyse- sample inputs from tissue to microbial samples. Collected samples do not require additional handling for processing and are ready for homogenization.