Learn More
Zymo Research Corporation DNA/RNA Shield Fecal Collection Tube (with Beads, 10 Pack)
SDP
Catalog No. NC2144346
Encompass_Preferred
DNA/RNA Shield Fecal Collection tubes are designed for the collection and preservation of nucleic acids from stool specimens. These fecal collection tubes take a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport. Samples stored in these fecal collection tubes are stable at ambient temperature, and can be frozen for longer-term storage. Simply collect a spoonful of the specimen into the stool collection tubes prefilled with preservative and shake vigorously to ensure proper stabilization. R1137 is pre-filled with 5 glass beads to help facilitate homogenization.
This item is not returnable. View return policy
Catalog No. NC2144346 Supplier Zymo Research Corporation Supplier No. R1137
DNA/RNA Shield Fecal Collection tubes are designed for the collection and preservation of nucleic acids from stool specimens. These fecal collection tubes take a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport. Samples stored in these fecal collection tubes are stable at ambient temperature, and can be frozen for longer-term storage. Simply collect a spoonful of the specimen into the stool collection tubes prefilled with preservative and shake vigorously to ensure proper stabilization. R1137 is pre-filled with 5 glass beads to help facilitate homogenization.