Learn More
Zymo Research Corporation DNA/RNA Shield Collection Tube w/ Swab (2ml fill) (50 pack)
SDP
Catalog No. 501251705
Encompass_Preferred
DNA/RNA Shield Collection tubes with swabs are a general purpose collection method for molecular-based applications. Many sample types can be collected including from the mouth, nose, throat, and wherever a swab may be deemed appropriate per application. To use, just swab any sample and break the tip into the collection device prefilled with the DNA/RNA Shield reagent. Samples are stabilized and ready for transport or storage.
Catalog No. 50-125-1705 Supplier Zymo Research Corporation Supplier No. R1109
DNA/RNA Shield Collection tubes with swabs are a general purpose collection method for molecular-based applications. Many sample types can be collected including from the mouth, nose, throat, and wherever a swab may be deemed appropriate per application. To use, just swab any sample and break the tip into the collection device prefilled with the DNA/RNA Shield reagent. Samples are stabilized and ready for transport or storage.