Learn More
LAMDA BIOTECH INC DNA SafeStain Plus
Supplier:  LAMDA BIOTECH INC C139
Encompass_Preferred
Description: DNA SafeStain Plus is a highly sensitive green fluorescent DNA/RNA staining reagent for detecting nucleic acids in agarose and polyacrylamide gels. This unique stain gives high sensitivity for detection of double-stranded or singlestranded DNA and RNA. Gels can be post-stained or the stain can be added to gels during gel casting or to the gel running buffer. DNA SafeStain Plus has two excitation wavelength peaks at about 290nm and 490nm, and an emission wavelength at 530nm, making it compatible with a standard UV light box, a blue-light transilluminator, or a gel reader equipped with visible light excitation; such as, a 488 nm laserbased gel scanner. DNA SafeStain Plus is in a 10,000X concentrated format that can be easily diluted 10,000 times for use in precast gel staining, or 5,000 times for use in post gel staining. Samples stained with DNA SafeStain Plus are compatible with downstream molecular biology applications; such as, gel extraction, and cloning
Catalog No. 50-204-5818