Learn More
Enzo Life Sciences AMPINEXT™ DNA Size Selection Kit (96Reactions)
SDP
Supplier:  Enzo Life Sciences ENZGEN5060096
Encompass_Preferred
The AMPINEXT™ DNA Size Selection Kit is a complete set of optimized reagents for quick removal of DNA fragments of <150 bps for library preparation in next generation sequencing applications. The kit utilizes magnetic bead technology and is suitable for Illumina, Life Technologies (SOLiD), Ion Torrent, and Roche/454 platforms. It can also be used for removing undesired larger DNA fragments by optimizing the bead to DNA volume ratio accordingly. The indicated number of reactions can be performed for a standard 50 µl solution input DNA sample. Assay Time: 30 minutes.
Catalog No. 50-201-3792