Learn More
Bioventures Inc DNA SIZING STANDARD
Catalog No. 50272360
Encompass
MAPMARKER, 70-400BP, TMR Labeled (70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400bp), 8fmoles/band/ul Concentration
Catalog No. 50-272-360 Supplier Bioventures Inc Supplier No. M1A
MAPMARKER, 70-400BP, TMR Labeled (70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400bp), 8fmoles/band/ul Concentration