Learn More
Bioventures Inc DNA SIZING STANDARD
Catalog No. 50272359
Encompass
MAPMARKER, 50-1000BP, TMR Labeled (50, 75, 100, 125, 150, 200, 250, 300, 350, 400, 450, 475, 500, 550, 600, 650, 700, 750, 800, 850, 900, 950, 1000bp), 16fmoles/band/ul Concentration
Catalog No. 50-272-359 Supplier Bioventures Inc Supplier No. M1
MAPMARKER, 50-1000BP, TMR Labeled (50, 75, 100, 125, 150, 200, 250, 300, 350, 400, 450, 475, 500, 550, 600, 650, 700, 750, 800, 850, 900, 950, 1000bp), 16fmoles/band/ul Concentration