Learn More
LAMDA BIOTECH INC DNA Spin Columns
Catalog No. 502045807
Encompass_Preferred
DNA Spin Columns with 2.0 ml collection tube. These columns are good for plasmid purification, genomic DNA isolation or DNA sample purification. They are the exactly same columns found in many different types of plasmid/DNA purification kits on the market.
Catalog No. 50-204-5807 Supplier LAMDA BIOTECH INC Supplier No. DNASC
DNA Spin Columns with 2.0 ml collection tube. These columns are good for plasmid purification, genomic DNA isolation or DNA sample purification. They are the exactly same columns found in many different types of plasmid/DNA purification kits on the market.