Learn More
MilliporeSigma™ DNA Staining Kit (Feulgen)
Catalog No. M1079070001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
M1079070001 1 Set
1 options
Catalog No. M1079070001 Supplier MilliporeSigma™ Supplier No. 1.07907.0001
Aids in diagnosis and prognosis of malignant tumor
  • For determination of DNA in histological cytological specimens
  • Applied directly at cell culture monolayers, imprints (touch preparations), smears from fine needle aspiration biopsies (FNAB), smears from exfoliated cells
  • Cell nuclei stains red-violet; cytoplasm, background un-colored
TRUSTED_SUSTAINABILITY
Specifications
Packaging Fibre Carton
Quantity 1 Set
Storage Requirements 15° to 25°C