Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ DNA Standard Marker
SDPSCI_ED

Catalog No. S29023 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S29023 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29023 Supplier Edvotek™ Supplier No. 7501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

New Edvotek standard marker features better separation, easier band measurements, and no unused bands.

  • Sufficient for 20 gels
  • DNA Standard ladder sizes, in base pairs: 6751, 3652, 2827, 1568, 1118, 825, 630

Specifications

Content And Storage Room Temperature
Detection Method Fluorescence
Format Tube
Label or Dye Not Applicable
Final Product Type DNA
Sample Type DNA
Labeling Target DNA
Product Type DNA Stardard Marker
Quantity 20 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.