Learn More
Teknova DNA Suspension Buffer (10mM Tris, 0.1mM EDTA), pH 8.0 1000mL, RNase/DNase, PCR Grade, Sterile. 8 pack.
SDP
Catalog No. 502745952
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-274-5952 Supplier Teknova Supplier No. 2T12208PK
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.