Learn More
Teknova DNA Suspension Buffer, pH 8.0. 10mL Tube, 25 Tubes per Pack, DNase/RNase Tested, Sterile. 4 pack.
SDP
Catalog No. 502745993
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-274-5993 Supplier Teknova Supplier No. T12354PK
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.