Learn More
Teknova DNA Suspension Buffer, pH 8.0. DNase/RNase Tested, PCR Grade. 1mL Tubes, 50-Pack, Sterile.
SDP
Catalog No. 502387974
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-238-7974 Supplier Teknova Supplier No. T0221
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.