Learn More
Teknova DNA Suspension Buffer, RNAse DNAse Tested (10mM Tris, 0.1mM EDTA), pH 8.0. 500mL, Sterile 8 pack.
SDP
Catalog No. 502387873
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-238-7873 Supplier Teknova Supplier No. T02588PK
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.