Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Synthesis Collection Vial

Catalog No. 400048 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400048 10 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 400048 Supplier Thermo Scientific Supplier No. 400048

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

These glass vials are for the collection of cleaved oligonucleotides during DNA synthesis.

These glass vials are for the collection of cleaved oligonucleotides during DNA synthesis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature. Includes 10 glass vials.
Detection Method Sequencing
Format Vial
Sample Type DNA
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
Product Type DNA Synthesis Collection Vial
Quantity 10 each
Shipping Condition Room Temperature
Tube Type Collection Tube

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.