Learn More
Research Products International Corp DNA Wash Buffer, 6 ml
SDP
Catalog No. 501978686
Encompass
DNA Wash Buffer (concentrate) is designed to use as a column wash to remove salts and other contaminants prior to DNA elution. This ensures high purity DNA to be recovered. Ethanol must be added to the concentrate prior to use. Instructions for each DNA Wash Buffer (concentrate) size is listed on the bottle and within the Zymo Research protocols it is used in.
Catalog No. 50-197-8686 Supplier Research Products International Corp Supplier No. ZD400326
DNA Wash Buffer (concentrate) is designed to use as a column wash to remove salts and other contaminants prior to DNA elution. This ensures high purity DNA to be recovered. Ethanol must be added to the concentrate prior to use. Instructions for each DNA Wash Buffer (concentrate) size is listed on the bottle and within the Zymo Research protocols it is used in.