Learn More
Zymo Research Corporation DNA Wash Buffer (Concentrate), 6 ml
SDP
Catalog No. NC0408495
Encompass_Preferred
This ensures high purity DNA to be recovered. Ethanol must be added to the concentrate prior to use. Instructions for each DNA Wash Buffer (concentrate) size is listed on the bottle and within the Zymo Research protocols it is used in.
Catalog No. NC0408495 Supplier Zymo Research Corporation Supplier No. D400326
This ensures high purity DNA to be recovered. Ethanol must be added to the concentrate prior to use. Instructions for each DNA Wash Buffer (concentrate) size is listed on the bottle and within the Zymo Research protocols it is used in.