Learn More
Medchemexpress LLC DNA2 inhibitor C5 | 35973-25-2 | 98.1% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128729
Encompass_Preferred
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. This product can be used in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • IC50 of 20 μM
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Can be used in breast cancer and colorectal cancer research
Catalog No. 50-003-42979