Learn More
Medchemexpress LLC DNA2 inhibitor C5 | 35973-25-2 | 98.5% | 1 ML
SDP
Catalog No. 5000340522
Encompass_Preferred
DNA2 inhibitor C5 is a potent, competitive, and specific inhibitor of the DNA2 nuclease. It has an IC50 of 20 μM and effectively inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. This compound is suitable for use in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor.
  • IC50 of 20 μM against DNA2 nuclease.
  • Inhibits multiple activities of DNA2, including nuclease, DNA-dependent ATPase, helicase, and DNA-binding.
  • Applicable in breast cancer and colorectal cancer research.
  • Solid appearance, light yellow to yellow in color.
Catalog No. 50-003-40522 Supplier Medchemexpress LLC Supplier No. HY12872910MM/1ML
DNA2 inhibitor C5 is a potent, competitive, and specific inhibitor of the DNA2 nuclease. It has an IC50 of 20 μM and effectively inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. This compound is suitable for use in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor.
  • IC50 of 20 μM against DNA2 nuclease.
  • Inhibits multiple activities of DNA2, including nuclease, DNA-dependent ATPase, helicase, and DNA-binding.
  • Applicable in breast cancer and colorectal cancer research.
  • Solid appearance, light yellow to yellow in color.