Learn More
Medchemexpress LLC DNA2 inhibitor C5 | 35973-25-2 | 98.5% | 10 MG
SDP
Catalog No. 5000340446
Encompass_Preferred
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. This product can be used in breast cancer and colorectal cancer research. It is for research use only and not sold to patients.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Useful in breast cancer and colorectal cancer research
  • Exhibits strong synergistic effect in inhibiting MCF7 cell survival with MK4827
  • Shows synthetic lethality with deficiencies in FEN1, EXO1, or MRE11 genes in mouse model
  • IC50 of 20 μM for DNA2 nuclease
Catalog No. 50-003-40446 Supplier Medchemexpress LLC Supplier No. HY12872910MG
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. This product can be used in breast cancer and colorectal cancer research. It is for research use only and not sold to patients.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Useful in breast cancer and colorectal cancer research
  • Exhibits strong synergistic effect in inhibiting MCF7 cell survival with MK4827
  • Shows synthetic lethality with deficiencies in FEN1, EXO1, or MRE11 genes in mouse model
  • IC50 of 20 μM for DNA2 nuclease